Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zfh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057502.5 | NP_476850.1 | 2 - 3 | 617 | 61 | |
NM_170522.4 | NP_733401.3 | 2 - 3 | 617 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |