Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zfh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057502.5 | NP_476850.1 | 4 - 5 | 3330 | 62 | |
NM_170522.4 | NP_733401.3 | 4 - 5 | 3330 | 62 | ||
NM_170523.3 | NP_733402.1 | 2 - 3 | 2393 | 62 | ||
GenBank mRNA | BT003277.1 | - | 2 - 3 | 2393 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |