Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057525.3 | NP_476873.2 | 3 - 4 | 1093 | 90 | |
GenBank mRNA | BT025203.2 | - | 3 - 4 | 1093 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |