Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057587.4 | NP_476935.1 | 4 - 5 | 1366 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |