Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ChAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057656.3 | NP_477004.5 | 4 - 5 | 1644 | 65 | |
NM_206517.2 | NP_996239.2 | 4 - 5 | 1644 | 65 | ||
GenBank mRNA | M63724.2 | - | 4 - 5 | 1622 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |