Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057822.4 | NP_477170.1 | 3 - 4 | 383 | 74 | |
NM_134310.3 | NP_599137.1 | 2 - 3 | 571 | 74 | ||
GenBank mRNA | AB035354.1 | - | 2 - 3 | 283 | 74 | |
AY070528.1 | - | 2 - 3 | 534 | 74 | ||
AY113404.1 | - | 2 - 3 | 572 | 74 | ||
D84315.1 | - | 3 - 4 | 351 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |