Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vha13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300478.1 | NP_001287407.1 | 2 - 3 | 283 | 89 | |
NM_058089.5 | NP_477437.1 | 2 - 3 | 283 | 89 | ||
GenBank mRNA | AF143200.1 | - | 2 - 3 | 258 | 89 | |
AY075202.1 | - | 2 - 3 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |