Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079505.3 | NP_524229.1 | 2 - 3 | 1432 | 116 | |
GenBank mRNA | BT003618.1 | - | 2322 | 116 | ||
U97685.1 | - | 1 - 2 | 1340 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |