Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cno | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275342.1 | NP_001262271.1 | 3 - 4 | 835 | 71 | |
NM_001275344.1 | NP_001262273.1 | 8 - 9 | 2115 | 71 | ||
NM_001275345.1 | NP_001262274.1 | 8 - 9 | 2115 | 71 | ||
NM_001316565.1 | NP_001303494.1 | 8 - 9 | 2115 | 71 | ||
NM_079508.4 | NP_524232.2 | 8 - 9 | 2115 | 71 | ||
NM_169026.3 | NP_730892.2 | 8 - 9 | 2115 | 71 | ||
GenBank mRNA | BT044227.1 | - | 8 - 9 | 2084 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |