Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kkv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079509.3 | NP_524233.1 | 3 - 4 | 518 | 69 | |
NM_169052.3 | NP_730928.2 | 3 - 4 | 518 | 69 | ||
NM_206430.3 | NP_996152.1 | 3 - 4 | 518 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |