Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300254.1 | NP_001287183.1 | 7 - 8 | 2270 | 90 | |
NM_079515.3 | NP_524239.2 | 7 - 8 | 2572 | 90 | ||
NM_169080.3 | NP_730966.1 | 7 - 8 | 2590 | 90 | ||
GenBank mRNA | BT025208.2 | - | 7 - 8 | 2562 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |