Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275399.1 | NP_001262328.1 | 2 - 3 | 761 | 109 | |
NM_001275400.1 | NP_001262329.1 | 2 - 3 | 578 | 109 | ||
NM_001275403.1 | NP_001262332.1 | 2 - 3 | 761 | 109 | ||
NM_079526.3 | NP_524250.3 | 2 - 3 | 761 | 109 | ||
NM_169171.2 | NP_731132.1 | 2 - 3 | 578 | 109 | ||
GenBank mRNA | AF217960.1 | - | 2 - 3 | 734 | 109 | |
AF281145.1 | - | 2 - 3 | 761 | 109 | ||
BT149892.1 | - | 2 - 3 | 725 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |