Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alpha-Est5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260036.2 | NP_001246965.1 | 2 - 3 | 384 | 66 | |
NM_079541.4 | NP_524265.3 | 2 - 3 | 381 | 66 | ||
GenBank mRNA | AY058345.1 | - | 2 - 3 | 516 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |