Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275482.1 | NP_001262411.1 | 12 - 13 | 2625 | 71 | |
NM_001275483.1 | NP_001262412.1 | 12 - 13 | 2628 | 71 | ||
NM_001382200.1 | NP_001368998.1 | 5 - 6 | 1116 | 71 | ||
NM_079564.4 | NP_524288.3 | 12 - 13 | 3987 | 71 | ||
NM_169274.2 | NP_731341.1 | 11 - 12 | 2424 | 71 | ||
NM_169275.2 | NP_731342.1 | 9 - 10 | 1865 | 71 | ||
NM_169276.2 | NP_731343.1 | 5 - 6 | 1332 | 71 | ||
GenBank mRNA | BT003462.1 | - | 12 - 13 | 3985 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |