Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alpha-Man-IIa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275484.1 | NP_001262413.1 | 4 - 5 | 3328 | 80 | |
NM_079567.3 | NP_524291.2 | 4 - 5 | 3325 | 80 | ||
GenBank mRNA | AY119464.1 | - | 4 - 5 | 3258 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |