Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hyd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300333.1 | NP_001287262.1 | 5 - 6 | 1084 | 108 | |
NM_079572.4 | NP_524296.2 | 5 - 6 | 1084 | 108 | ||
GenBank mRNA | BT150484.1 | - | 5 - 6 | 1068 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |