Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irp-1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079579.3 | NP_524303.2 | 2 - 3 | 157 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |