Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irp-1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079579.3 | NP_524303.2 | 6 - 7 | 1147 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |