Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TkR86C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104271.1 | NP_001097741.1 | 2 - 3 | 1283 | 110 | |
NM_079580.3 | NP_524304.2 | 2 - 3 | 872 | 110 | ||
GenBank mRNA | BT029262.1 | - | 2 - 3 | 1283 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Drosophila melanogaster |