Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TkR86C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079580.3 | NP_524304.2 | 7 - 8 | 1766 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Drosophila melanogaster |