Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RpL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079592.3 | NP_524316.1 | 5 - 6 | 925 | 88 | |
NM_169378.2 | NP_731548.2 | 5 - 6 | 925 | 88 | ||
NM_169380.2 | NP_731550.2 | 5 - 6 | 1232 | 88 | ||
GenBank mRNA | AF016835.1 | - | 5 - 6 | 922 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |