Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079607.3 | NP_524331.2 | 3 - 4 | 366 | 69 | |
GenBank mRNA | AY070666.1 | - | 3 - 4 | 373 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |