Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yellow-f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079611.2 | NP_524335.1 | 1 - 2 | 225 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |