Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: His4r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275647.1 | NP_001262576.1 | 2 - 3 | 238 | 87 | |
NM_079628.4 | NP_524352.1 | 1 - 2 | 448 | 87 | ||
NM_169592.3 | NP_731927.1 | 2 - 3 | 271 | 87 | ||
NM_169593.3 | NP_731928.1 | 2 - 3 | 340 | 87 | ||
GenBank mRNA | BT001842.1 | - | 2 - 3 | 286 | 87 | |
X97438.1 | - | 232 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |