Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202301.2 | NP_001189230.1 | 4 - 5 | 1602 | 123 | |
NM_079649.4 | NP_524373.1 | 4 - 5 | 1602 | 123 | ||
GenBank mRNA | AF071502.1 | - | 4 - 5 | 1501 | 123 | |
BT031130.1 | - | 4 - 5 | 1547 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |