Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202301.2 | NP_001189230.1 | 7 - 8 | 3408 | 58 | |
NM_079649.4 | NP_524373.1 | 7 - 8 | 3600 | 58 | ||
GenBank mRNA | AF071502.1 | - | 7 - 8 | 3499 | 58 | |
BT031130.1 | - | 7 - 8 | 3545 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |