Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079657.4 | NP_524381.1 | 2 - 3 | 262 | 100 | |
GenBank mRNA | AF014952.1 | - | 1 - 2 | 85 | 100 | |
AF093240.1 | - | 2 - 3 | 231 | 100 | ||
AY089680.1 | - | 2 - 3 | 262 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |