Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cher | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202309.1 | NP_001189238.1 | 5 - 6 | 1222 | 75 | |
NM_001275728.1 | NP_001262657.2 | 6 - 7 | 1532 | 75 | ||
NM_001275729.1 | NP_001262658.1 | 5 - 6 | 1222 | 75 | ||
NM_001275730.1 | NP_001262659.2 | 7 - 8 | 1547 | 75 | ||
NM_079659.4 | NP_524383.3 | 5 - 6 | 1283 | 75 | ||
NM_206502.3 | NP_996224.2 | 5 - 6 | 1283 | 75 | ||
GenBank mRNA | AF188360.1 | - | 5 - 6 | 1259 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |