Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cher | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202309.1 | NP_001189238.1 | 9 - 10 | 4156 | 92 | |
NM_001275728.1 | NP_001262657.2 | 11 - 12 | 5033 | 92 | ||
NM_001275729.1 | NP_001262658.1 | 10 - 11 | 4723 | 92 | ||
NM_001275730.1 | NP_001262659.2 | 12 - 13 | 5048 | 92 | ||
NM_079659.4 | NP_524383.3 | 9 - 10 | 4217 | 92 | ||
NM_206502.3 | NP_996224.2 | 10 - 11 | 4784 | 92 | ||
GenBank mRNA | AF188360.1 | - | 9 - 10 | 4193 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |