Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: beat-IIa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300446.1 | NP_001287375.1 | 3 - 4 | 1290 | 71 | |
NM_079660.4 | NP_524384.3 | 4 - 5 | 1445 | 71 | ||
GenBank mRNA | AF325713.1 | - | 4 - 5 | 1244 | 71 | |
AY118373.1 | - | 4 - 5 | 1443 | 71 | ||
BT023786.1 | - | 3 - 4 | 541 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |