Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: beat-IIa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300446.1 | NP_001287375.1 | 4 - 5 | 1416 | 90 | |
NM_079660.4 | NP_524384.3 | 5 - 6 | 1571 | 90 | ||
GenBank mRNA | AF325713.1 | - | 5 - 6 | 1370 | 90 | |
AY118373.1 | - | 5 - 6 | 1569 | 90 | ||
BT023786.1 | - | 4 - 5 | 667 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |