Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260223.1 | NP_001247152.1 | 2 - 3 | 557 | 96 | |
NM_079661.5 | NP_524385.2 | 1 - 2 | 500 | 96 | ||
NM_169756.2 | NP_732229.1 | 2 - 3 | 562 | 96 | ||
GenBank mRNA | AY113353.1 | - | 2 - 3 | 505 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |