Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260223.1 | NP_001247152.1 | 3 - 4 | 631 | 66 | |
NM_079661.5 | NP_524385.2 | 2 - 3 | 574 | 66 | ||
NM_169756.2 | NP_732229.1 | 3 - 4 | 636 | 66 | ||
GenBank mRNA | AY113353.1 | - | 3 - 4 | 579 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |