Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275739.1 | NP_001262668.1 | 3 - 4 | 293 | 76 | |
NM_079662.3 | NP_524386.1 | 3 - 4 | 297 | 76 | ||
GenBank mRNA | BT023233.1 | - | 3 - 4 | 274 | 76 | |
X72302.1 | - | 295 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |