Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275764.1 | NP_001262693.1 | 3 - 4 | 1733 | 67 | |
NM_079671.2 | NP_524395.2 | 3 - 4 | 1733 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |