Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sqz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079679.3 | NP_524403.1 | 2 - 3 | 1254 | 73 | |
GenBank mRNA | AF173847.1 | - | 2 - 3 | 1228 | 73 | |
BT150373.1 | - | 2 - 3 | 1244 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |