Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300473.1 | NP_001287402.1 | 6 - 7 | 509 | 70 | |
NM_079680.2 | NP_524404.1 | 6 - 7 | 602 | 70 | ||
GenBank mRNA | AF246993.1 | - | 6 - 7 | 567 | 70 | |
BT083455.1 | - | 6 - 7 | 509 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |