Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dys | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043257.2 | NP_001036722.1 | 11 - 12 | 6723 | 125 | |
NM_001043258.2 | NP_001036723.1 | 11 - 12 | 6723 | 125 | ||
NM_001043259.2 | NP_001036724.1 | 10 - 11 | 5724 | 125 | ||
NM_001043261.2 | NP_001036726.1 | 10 - 11 | 6436 | 125 | ||
NM_001043263.2 | NP_001036728.1 | 10 - 11 | 6650 | 125 | ||
NM_001275801.1 | NP_001262730.1 | 9 - 10 | 5609 | 125 | ||
NM_001275803.1 | NP_001262732.1 | 10 - 11 | 5724 | 125 | ||
GenBank mRNA | AF277386.1 | - | 9 - 10 | 5293 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |