Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dys | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043256.2 | NP_001036721.1 | 3 - 4 | 1813 | 83 | |
NM_001043257.2 | NP_001036722.1 | 19 - 20 | 7660 | 83 | ||
NM_001043258.2 | NP_001036723.1 | 19 - 20 | 7660 | 83 | ||
NM_001043259.2 | NP_001036724.1 | 17 - 18 | 6592 | 83 | ||
NM_001043261.2 | NP_001036726.1 | 18 - 19 | 7373 | 83 | ||
NM_001043263.2 | NP_001036728.1 | 18 - 19 | 7587 | 83 | ||
NM_001275801.1 | NP_001262730.1 | 16 - 17 | 6477 | 83 | ||
NM_001275803.1 | NP_001262732.1 | 17 - 18 | 6592 | 83 | ||
NM_001275804.1 | NP_001262733.1 | 7 - 8 | 1135 | 83 | ||
GenBank mRNA | AF277386.1 | - | 16 - 17 | 6161 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |