Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dys | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043256.2 | NP_001036721.1 | 4 - 5 | 1952 | 109 | |
NM_001043257.2 | NP_001036722.1 | 20 - 21 | 7799 | 109 | ||
NM_001043258.2 | NP_001036723.1 | 20 - 21 | 7799 | 109 | ||
NM_001043259.2 | NP_001036724.1 | 18 - 19 | 6731 | 109 | ||
NM_001043261.2 | NP_001036726.1 | 19 - 20 | 7512 | 109 | ||
NM_001043263.2 | NP_001036728.1 | 19 - 20 | 7726 | 109 | ||
NM_001275801.1 | NP_001262730.1 | 17 - 18 | 6616 | 109 | ||
NM_001275803.1 | NP_001262732.1 | 18 - 19 | 6731 | 109 | ||
NM_001275804.1 | NP_001262733.1 | 8 - 9 | 1274 | 109 | ||
GenBank mRNA | AF277386.1 | - | 17 - 18 | 6300 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |