Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dys | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043256.2 | NP_001036721.1 | 7 - 8 | 2296 | 90 | |
NM_001043257.2 | NP_001036722.1 | 23 - 24 | 8143 | 90 | ||
NM_001043258.2 | NP_001036723.1 | 23 - 24 | 8143 | 90 | ||
NM_001043259.2 | NP_001036724.1 | 21 - 22 | 7075 | 90 | ||
NM_001043260.2 | NP_001036725.1 | 3 - 4 | 3079 | 90 | ||
NM_001043261.2 | NP_001036726.1 | 22 - 23 | 7856 | 90 | ||
NM_001043263.2 | NP_001036728.1 | 22 - 23 | 8070 | 90 | ||
NM_001275801.1 | NP_001262730.1 | 20 - 21 | 6960 | 90 | ||
NM_001275803.1 | NP_001262732.1 | 21 - 22 | 7075 | 90 | ||
NM_001275804.1 | NP_001262733.1 | 11 - 12 | 1618 | 90 | ||
GenBank mRNA | AF277386.1 | - | 20 - 21 | 6644 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |