Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or92a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079690.2 | NP_524414.2 | 1 - 2 | 817 | 95 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Drosophila melanogaster |