Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260283.2 | NP_001247212.1 | 9 - 10 | 3088 | 72 | |
NM_001260284.2 | NP_001247213.1 | 8 - 9 | 2943 | 72 | ||
NM_001260285.2 | NP_001247214.1 | 8 - 9 | 2895 | 72 | ||
NM_001260286.2 | NP_001247215.1 | 9 - 10 | 3806 | 72 | ||
NM_001300505.1 | NP_001287434.1 | 8 - 9 | 3048 | 72 | ||
NM_001300506.1 | NP_001287435.1 | 8 - 9 | 4462 | 72 | ||
NM_079699.4 | NP_524423.2 | 7 - 8 | 3737 | 72 | ||
NM_169940.4 | NP_732576.1 | 8 - 9 | 3140 | 72 | ||
NM_169941.2 | NP_732577.1 | 7 - 8 | 2982 | 72 | ||
GenBank mRNA | AY118680.1 | - | 8 - 9 | 3086 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |