Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mvl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170197.1 | NP_001163668.1 | 10 - 11 | 3925 | 96 | |
NM_001275866.1 | NP_001262795.1 | 11 - 12 | 1796 | 96 | ||
NM_079701.5 | NP_524425.2 | 11 - 12 | 2112 | 96 | ||
NM_169944.3 | NP_732584.1 | 11 - 12 | 1796 | 96 | ||
GenBank mRNA | AY058543.1 | - | 11 - 12 | 2071 | 96 | |
BT044567.1 | - | 10 - 11 | 3923 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |