Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: InR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144621.3 | NP_001138093.1 | 10 - 11 | 4070 | 72 | |
NM_001144622.2 | NP_001138094.1 | 9 - 10 | 3371 | 72 | ||
NM_001144623.2 | NP_001138095.1 | 9 - 10 | 3437 | 72 | ||
NM_079712.6 | NP_524436.2 | 10 - 11 | 4080 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |