Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079730.3 | NP_524454.2 | 10 - 11 | 2213 | 71 | |
GenBank mRNA | AY060363.1 | - | 10 - 11 | 2195 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |