Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079730.3 | NP_524454.2 | 9 - 10 | 2039 | 56 | |
GenBank mRNA | AY060363.1 | - | 9 - 10 | 2021 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |