Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cont | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141204.4 | NP_649461.2 | 4 - 5 | 3011 | 78 | |
GenBank mRNA | AY229991.1 | - | 3 - 4 | 2734 | 78 | |
BT021333.1 | - | 4 - 5 | 3007 | 78 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |