Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eIF3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141213.4 | NP_649470.2 | 3 - 4 | 3524 | 64 | |
NM_168999.3 | NP_730838.3 | 3 - 4 | 3524 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster Inventoried |