Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141227.3 | NP_649484.3 | 2 | 871 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |